missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-oxocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 10702105 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702105 5 g Glass bottle 5g
10077102 250 mg Glass bottle 250mg
10670835 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702105 Supplier Thermo Scientific Acros Supplier No. 379910050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17159-79-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00013285
InChI Key ZXYAWONOWHSQRU-UHFFFAOYSA-N
Synonym ethyl 4-oxocyclohexanecarboxylate, ethyl 4-cyclohexanonecarboxylate, 4-ethoxycarbonyl cyclohexanone, ethyl cyclohexanone-4-carboxylate, ethyl-4-oxo-cyclohexane carboxylate, 4-oxo-cyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-oxo-, ethyl ester, ethyl4-oxocyclohexanecarboxylate, 4-cyclohexanonecarboxylic acid ethyl ester, ethyl 4-oxocyclohexane carboxylate
PubChem CID 317638
IUPAC Name ethyl 4-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(=O)CC1

Specifications

Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 150°C to 152°C (40.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (O=)C6H9CO2C2H5
Refractive Index 1.4580 to 1.461
Quantity 5 g
Specific Gravity 1.076
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4-oxocyclohexanecarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.