missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Oxo-4-(3-pyridyl)butanoate, TRC

Product Code. 30380280
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380280 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380280 Supplier Toronto Research Chemicals Supplier No. TRCE925400250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-Oxo-4-(3-pyridyl)butanoate
CAS 59086-27-0
Molecular Formula C11 H13 N O3
Quantity 250 mg
Synonym 3-Pyridinebutanoic acid, γ-oxo-, ethyl ester, Ethyl γ-oxo-3-pyridinebutanoate, Ethyl 3-nicotinoyl propionate, Ethyl 4-oxo-4-(3-pyridyl)butanoate, Ethyl 4-oxo-4-(pyridin-3-yl)butanoate, 4-Oxo-4-(3-pyridyl)butanoic acid ethyl ester, γ-Oxo-3-pyridinebutanoic acid ethyl ester
InChI Formula InChI=1S/C11H13NO3/c1-2-15-11(14)6-5-10(13)9-4-3-7-12-8-9/h3-4,7-8H,2,5-6H2,1H3
SMILES CCOC(=O)CCC(=O)c1cccnc1
IUPAC Name ethyl 4-oxo-4-pyridin-3-ylbutanoate
Molecular Weight (g/mol) 207.23
Formula Weight 207.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.