missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl [(4-Methylphenyl)sulphonyl]carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ethyl [(4-Methylphenyl)sulphonyl]carbamate
Specifications
Specifications
| Chemical Name or Material | Ethyl [(4-Methylphenyl)sulphonyl]carbamate |
| CAS | 5577-13-9 |
| Analyte or Component Names | Ethyl [(4-Methylphenyl)sulphonyl]carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 N O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl [(4-Methylphenyl)sulphonyl]carbamate, Gliclazide Imp. C (EP), Gliclazide Impurity C |
| InChI Formula | InChI=1S/C10H13NO4S/c1-3-15-10(12)11-16(13,14)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3,(H,11,12) |
| SMILES | CCOC(=O)NS(=O)(=O)c1ccc(C)cc1 |
| IUPAC Name | ethyl N-(4-methylphenyl)sulfonylcarbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction