missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-5-imidazolecarboxylate, 98%

Product Code. 10343672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10343672 10 g Glass bottle 10g
10353672 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10343672 Supplier Thermo Scientific Acros Supplier No. 190340100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51605-32-4 | C7H10N2O2 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51605-32-4
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00005199
InChI Key VLDUBDZWWNLZCU-UHFFFAOYSA-N
Synonym ethyl 4-methyl-5-imidazolecarboxylate, ethyl 4-methyl-1h-imidazole-5-carboxylate, ethyl-4-methyl-5-imidazolecarboxylate, 1h-imidazole-5-carboxylic acid, 4-methyl-, ethyl ester, 1h-imidazole-4-carboxylic acid, 5-methyl-, ethyl ester, 4-carboethoxy-5-methylimidazole, ethyl 4-methylimidazole-5-carboxylate, ethyl 5-methyl-4-imidazolecarboxylate, 5-methyl-3h-imidazole-4-carboxylic acid ethyl ester, ethyl 5-methylimidazole-4-carboxylate
PubChem CID 99200
IUPAC Name ethyl 5-methyl-1H-imidazole-4-carboxylate
SMILES CCOC(=O)C1=C(C)NC=N1

Specifications

Melting Point 204°C to 206°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 25, I, 534
Solubility Information Solubility in water: insoluble in water
Formula Weight 154.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.