missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxyphenylacetate, TRC

Product Code. 30010489
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010489 25 g 25g
30003271 1 g 1g
30012524 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010489 Supplier Toronto Research Chemicals Supplier No. TRCE91908025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17138-28-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
Synonym 4-Hydroxyphenylacetic Acid Ethyl Ester, 4-Hydroxybenzeneacetic Acid Ethyl Ester, (p-Hydroxyphenyl)acetic Acid Ethyl Ester, 2-(4-Hydroxyphenyl)acetic Acid Ethyl Ester, Ethyl (p-Hydroxyphenyl)acetate, Ethyl 2-(4-Hydroxyphenyl)acetate, Ethyl 2-(p-Hydroxyphenyl)acetate;
IUPAC Name ethyl 2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)Cc1ccc(O)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H12O3/c1-2-13-10(12)7-8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3
Formula Weight 180.0786
Chemical Name or Material Ethyl 4-Hydroxyphenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.