missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%

Product Code. p-7028324
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11461080

Brand: Thermo Scientific Alfa Aesar B25572.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17159-80-7 | C9H16O3 | 172.22 g/mol

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17159-80-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00192156,MFCD16295187
InChI Key BZKQJSLASWRDNE-UHFFFAOYSA-N
Synonym ethyl 4-hydroxycyclohexanecarboxylate, trans-ethyl 4-hydroxycyclohexanecarboxylate, ethyl trans-4-hydroxycyclohexanecarboxylate, 4-hydroxy-cyclohexanecarboxylic acid ethyl ester, ethyl 4-hydroxycyclohexane carboxylate, cis-ethyl 4-hydroxycyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, 4-hydroxycyclohexanecarboxylic acid ethyl ester, trans-4-hydroxycyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, cis
PubChem CID 86973
IUPAC Name ethyl 4-hydroxycyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(O)CC1

Specifications

Density 1.068
Boiling Point 127°C to 134°C (0.1 mmHg)
Flash Point >110(230°F)
Refractive Index 1.467
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.