missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-formylpyrrole-2-carboxylate, 96%

Product Code. p-7041702
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15432129

Brand: Thermo Scientific Alfa Aesar H64776.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7126-57-0 | C8H9NO3 | 167.164 g/mol

Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

Solubility
Miscible with ethyl acetate.

Notes
Incompatible with steel and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7126-57-0
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD04122505
InChI Key WVSAWXIWWNJTAV-UHFFFAOYSA-N
Synonym ethyl 4-formylpyrrole-2-carboxylate, 4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-formyl-, ethyl ester, acmc-20akyk, pubchem14845, ethyl4-formyl-1h-pyrrole-2-carboxylate, ethyl 4-methanoyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-, ethyl ester
PubChem CID 321486
IUPAC Name ethyl 4-formyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(=CN1)C=O

Specifications

Melting Point 101°C to 105°C
Quantity 250 mg
Beilstein 473842
Solubility Information Miscible with ethyl acetate.
Formula Weight 167.16
Percent Purity 96%
Chemical Name or Material Ethyl 4-formylpyrrole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.