missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-formylpyrrole-2-carboxylate, 96%

Product Code. 15422129
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422129 5 g 5g
15432129 250 mg 250mg
15412129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422129 Supplier Thermo Scientific Alfa Aesar Supplier No. H64776.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7126-57-0 | C8H9NO3 | 167.164 g/mol

Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

Solubility
Miscible with ethyl acetate.

Notes
Incompatible with steel and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7126-57-0
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD04122505
InChI Key WVSAWXIWWNJTAV-UHFFFAOYSA-N
Synonym ethyl 4-formylpyrrole-2-carboxylate, 4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-formyl-, ethyl ester, acmc-20akyk, pubchem14845, ethyl4-formyl-1h-pyrrole-2-carboxylate, ethyl 4-methanoyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-, ethyl ester
PubChem CID 321486
IUPAC Name ethyl 4-formyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(=CN1)C=O

Specifications

Melting Point 101°C to 105°C
Quantity 5 g
Beilstein 473842
Solubility Information Miscible with ethyl acetate.
Formula Weight 167.16
Percent Purity 96%
Chemical Name or Material Ethyl 4-formylpyrrole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.