missing translation for 'onlineSavingsMsg'
Learn More

ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate, TRC

Product Code. 30374354
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374354 500 mg
30369709 50 mg
30370460 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374354 Supplier Toronto Research Chemicals Supplier No. TRCE903033500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-64-6
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
Synonym Pyrrole-2-carboxylic acid, 4-formyl-3,5-dimethyl-, ethyl ester (6CI,7CI,8CI), Ethyl 4-formyl-3,5-dimethylpyrrole-2-carboxylate, NSC 13665
SMILES CCOC(=O)c1[nH]c(C)c(C=O)c1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H13NO3/c1-4-14-10(13)9-6(2)8(5-12)7(3)11-9/h5,11H,4H2,1-3H3
Formula Weight 195.0895
Chemical Name or Material ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.