missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-fluorobenzoate, 99%

Product Code. p-7025806
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11457024

Brand: Thermo Scientific Alfa Aesar B21484.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 451-46-7 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-46-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00000351
InChI Key UMPRJGKLMUDRHL-UHFFFAOYSA-N
Synonym 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
PubChem CID 67976
IUPAC Name ethyl 4-fluorobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 25°C to 27°C
Density 1.146
Boiling Point 209°C to 210°C
Flash Point 81°C (177°F)
Odor Recognizable
Refractive Index 1.487
Quantity 5 g
Beilstein 1866794
Merck Index 14,4171
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.