missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Ethoxybenzoate, TRC

Product Code. 30137987
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137987 50 g 50g
30137308 10 g 10g
30003733 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137987 Supplier Toronto Research Chemicals Supplier No. TRCE93756550G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23676-09-7
Molecular Formula C11 H14 O3
Molecular Weight (g/mol) 194.23
Synonym Benzoic acid, p-ethoxy-, ethyl ester (6CI,7CI,8CI), 4-Ethoxy ethylbenzoate, 4-Ethoxybenzoic acid ethyl ester, Ethyl 4-ethoxybenzoate, Ethyl p-ethoxybenzoate, NSC 405367, p-Ethoxyethyl benzoate, Benzoic acid, 4-ethoxy-, ethyl ester, 4-Ethoxybenzoic Acid-Ethyl Ester
IUPAC Name ethyl 4-ethoxybenzoate
SMILES CCOC(=O)c1ccc(OCC)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C11H14O3/c1-3-13-10-7-5-9(6-8-10)11(12)14-4-2/h5-8H,3-4H2,1-2H3
Formula Weight 194.0943
Chemical Name or Material 4-Ethoxybenzoic Acid-Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.