missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-dimethylaminobenzoate, 99+%

Product Code. 10529600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10529600 5 g Glass Bottle 5g
10417690 100 g Plastic bottle 100g
10793465 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529600 Supplier Thermo Scientific Acros Supplier No. 118400050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10287-53-3 | C11H15NO2 | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10287-53-3
MDL Number MFCD00009115
InChI Key FZUGPQWGEGAKET-UHFFFAOYSA-N
Synonym ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x
PubChem CID 25127
ChEBI CHEBI:52073
IUPAC Name ethyl 4-(dimethylamino)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 62°C to 65°C
Color White to Yellow
Density 1.0600g/mL
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Molecular Formula C11H15NO2
Linear Formula (CH3)2NC6H4CO2C2H5
Quantity 5 g
Beilstein 14, I, 571
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: >50g/100 mL acetone (20 c),18g/100mL ethanol (20 c),2g/100mL mineral oil (20 c)
Molecular Weight (g/mol) 193.25
Formula Weight 193.25
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-dimethylaminobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-dimethylaminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.