missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorophenylacetate, 98+%

Product Code. 10698814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10698814 5 g Glass bottle 5g
10269243 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10698814 Supplier Thermo Scientific Acros Supplier No. 388840050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14062-24-9 | C10H11ClO2 | 198.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-24-9
MDL Number MFCD00018791
InChI Key UTWBWFXECVFDPZ-UHFFFAOYSA-N
Synonym ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p
PubChem CID 84177
IUPAC Name ethyl 2-(4-chlorophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)Cl

Specifications

Color Colorless
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Low Melting Solid
Assay Percent Range 98+%
Molecular Formula C10H11ClO2
Linear Formula ClC6H4CH2C(O)OCH2CH3
Molecular Weight (g/mol) 198.646
Formula Weight 198.65
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-chlorophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-chlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.