missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorobutyrate, 98%

Product Code. 11494300
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494300 100 g 100g
11404310 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494300 Supplier Thermo Scientific Alfa Aesar Supplier No. A11070.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3153-36-4 | C6H11ClO2 | 150.602 g/mol

Ethyl 4-chlorobutyrate is used in organic synthesis. And also used for producting 4-Iodo-butyric acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chlorobutyrate is used in organic synthesis. And also used for producting 4-Iodo-butyric acid ethyl ester.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-36-4
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001004
InChI Key OPXNFHAILOHHFO-UHFFFAOYSA-N
Synonym ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
PubChem CID 18474
IUPAC Name ethyl 4-chlorobutanoate
SMILES CCOC(=O)CCCCl

Specifications

Density 1.077
Boiling Point 185°C to 186°C
Flash Point 76°C (169°F)
Refractive Index 1.432
Quantity 100 g
Beilstein 1098810
Solubility Information Insoluble in water.
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material Ethyl 4-chlorobutyrate
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-chlorobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl 4-chlorobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.