missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorobenzoate, 98+%

Product Code. 11494214
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11494214

Brand: Thermo Scientific Alfa Aesar A12540.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7335-27-5 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7335-27-5
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00013645
InChI Key RWBYCMPOFNRISR-UHFFFAOYSA-N
Synonym ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705
PubChem CID 81785
IUPAC Name ethyl 4-chlorobenzoate
SMILES CCOC(=O)C1=CC=C(Cl)C=C1

Specifications

Density 1.18
Boiling Point 237°C to 239°C
Refractive Index 1.524
Quantity 50 g
Beilstein 2045882
Formula Weight 184.62
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-chlorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.