missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloroacetoacetate, 98%

Product Code. 10183760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183760 250 g Glass bottle 250g
10022380 50 g Glass bottle 50g
11995101 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183760 Supplier Thermo Scientific Acros Supplier No. 177642500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-07-3 | C6H9ClO3 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-07-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000939
InChI Key OHLRLMWUFVDREV-UHFFFAOYSA-N
Synonym ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
PubChem CID 69484
IUPAC Name ethyl 4-chloro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)CCl

Specifications

Melting Point -8.0°C
Color Colorless to Red-Yellow
Density 1.2100g/mL
Boiling Point 103.0°C (16.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2COCH2CO2C2H5
Refractive Index 1.4510 to 1.4530
Quantity 250 g
Beilstein 03,663
Specific Gravity 1.21
Solubility Information Solubility in water: 47.5g/L water (20°C). Other solubilities: soluble in ether,benzene,chloroform,,methylenechloride and carbontetrachloride
Formula Weight 164.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4-chloroacetoacetate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-chloroacetoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.