missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloroacetoacetate, 98%, Thermo Scientific Chemicals

Product Code. 11995101
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-07-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000939
InChI Key OHLRLMWUFVDREV-UHFFFAOYSA-N
Synonym ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
PubChem CID 69484
IUPAC Name ethyl 4-chloro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)CCl

Specifications

Melting Point -8.0°C
Color Colorless to Red-Yellow
Density 1.2100g/mL
Boiling Point 103.0°C (16.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2COCH2CO2C2H5
Refractive Index 1.4510 to 1.4530
Quantity 1 kg
Beilstein 03,663
Specific Gravity 1.21
Solubility Information Solubility in water: 47.5g/L water (20°C). Other solubilities: soluble in ether,benzene,chloroform,,methylenechloride and carbontetrachloride
Formula Weight 164.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4-chloroacetoacetate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-chloroacetoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'