missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloroacetoacetate, 97%

Product Code. 11447547
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447547 50 g 50g
11457547 250 g 250g
11437547 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447547 Supplier Thermo Scientific Alfa Aesar Supplier No. A15641.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-07-3 | C6H9ClO3 | 164.585 g/mol

Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-chloroacetoacetate is used as a precursor in the preparation of phosphorous ylides. It finds application in regular film coating, pharmaceutical coating and formulation.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with strong acids, strong bases, reducing agents and strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-07-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00000939
InChI Key OHLRLMWUFVDREV-UHFFFAOYSA-N
Synonym ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate
PubChem CID 69484
IUPAC Name ethyl 4-chloro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)CCl

Specifications

Melting Point -8°C
Density 1.212
Boiling Point 220°C
Flash Point 92°C (198°F)
Refractive Index 1.452
Quantity 50 g
UN Number UN2922
Beilstein 1761275
Solubility Information Miscible with water and most organic solvents.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloroacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.