missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%, Thermo Scientific™

Product Code. 15512303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15512303 5 g 5g
15502303 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15512303 Supplier Thermo Scientific Acros Supplier No. 449680050
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 77173-67-2
Molecular Formula C13H9ClN2O2
Molecular Weight (g/mol) 260.68
InChI Key MHOFAUCLSGVKOH-UHFFFAOYSA-N
Synonym ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
PubChem CID 3761179
IUPAC Name ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 260.68
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
compliance-icons
Product Identifier
  • Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.