missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 97%, Thermo Scientific™

Product Code. 11375599
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375599 250 mg 250mg
11385599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375599 Supplier Thermo Scientific Alfa Aesar Supplier No. H50515.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22931-71-1
Molecular Formula C13H12ClNO3
Molecular Weight (g/mol) 265.693
MDL Number MFCD00173417
InChI Key QURGQUFEJWWDRF-UHFFFAOYSA-N
Synonym gnf-pf-3551, 4-chloro-6-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-6-methoxy-3-quinolinecarboxylate, acmc-20a6z6, 3-carbethoxy-4-chloro-6-methoxyquinoline, 4-chloro-3-ethoxycarbonyl-6-methoxyquinoline, 6-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4chloro-6methoxyquinoline-3-carboxylate, ethyl 4-chloro-6-methoxyquinolin-3-carboxylate
PubChem CID 304558
IUPAC Name ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)OC

Specifications

Melting Point 88°C to 90°C
Quantity 250 mg
Formula Weight 265.7
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.