missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-hydroxy-5-nitronicotinate, 97%, Thermo Scientific™

Product Code. 15405987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210835-74-7
Molecular Formula C8H7ClN2O5
Molecular Weight (g/mol) 246.603
MDL Number MFCD22989339
InChI Key FETZVQJUBMRFJE-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, ethyl 4-chloro-6-hydroxy-5-nitronicotinate, ethyl 4-chloro-6-hydroxy-5-nitropyridine-3-carboxylate, ethylchloronitrooxodihydropyridinecarboxylate, 4-chloro-6-hydroxy-5-nitronicotinic acid ethyl ester, ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 4-chloro-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester
PubChem CID 59225818
IUPAC Name ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
SMILES CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 176°C to 178°C
Color Pink to White
Quantity 1 g
Formula Weight 246.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-chloro-6-hydroxy-5-nitronicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'