missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-3-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 11483937
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483937 5 g 5g
11493937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483937 Supplier Thermo Scientific Alfa Aesar Supplier No. A19338.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16588-16-2
Molecular Formula C9H8ClNO4
Molecular Weight (g/mol) 229.616
MDL Number MFCD00219621
InChI Key BLNLZRQIUGDTAO-UHFFFAOYSA-N
Synonym benzoic acid, 4-chloro-3-nitro-, ethyl ester, 4-chloro-3-nitrobenzoic acid ethyl ester, 4-chloro-3-nitro-benzoic acid ethyl ester, ethyl 4-chloranyl-3-nitro-benzoate, acmc-20a7tu, ethyl4-chloro-3-nitrobenzoate, ethyl 4-chloro-3-nitro-benzoate, ethyl-4-chloro-3-nitro-benzoate, ethyl-4-chloro-3-nitrobenzoate, 3-nitro-4-chlorobenzoic acid ethyl ester
PubChem CID 1268247
IUPAC Name ethyl 4-chloro-3-nitrobenzoate
SMILES CCOC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 53°C to 55°C
Quantity 5 g
Formula Weight 229.62
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.