missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%

Product Code. 11450567
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450567 5 g 5g
11460567 25 g 25g
11470567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450567 Supplier Thermo Scientific Alfa Aesar Supplier No. B22429.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5909-24-0 | C8H9ClN2O2S | 232.682 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5909-24-0
Molecular Formula C8H9ClN2O2S
Molecular Weight (g/mol) 232.682
MDL Number MFCD00006085
InChI Key SNNHLSHDDGJVDM-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester
PubChem CID 80008
IUPAC Name ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)SC

Specifications

Melting Point 60°C to 63°C
Quantity 5 g
Formula Weight 232.69
Percent Purity 98%
Chemical Name or Material Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.