missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromocrotonate, tech. 80%

Product Code. 11484183
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484183 250 g 250g
11464183 10 g 10g
11474183 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484183 Supplier Thermo Scientific Alfa Aesar Supplier No. A10371.30
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37746-78-4 | C6H9BrO2 | 193.04 g/mol

Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with acids, bases, oxidizing agents, reducing agents. Protect from heat. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37746-78-4
Molecular Formula C6H9BrO2
Molecular Weight (g/mol) 193.04
MDL Number MFCD00000247
InChI Key FHGRPBSDPBRTLS-ONEGZZNKSA-N
Synonym ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate
PubChem CID 5373944
IUPAC Name ethyl (E)-4-bromobut-2-enoate
SMILES CCOC(=O)C=CCBr

Specifications

Density 1.402
Boiling Point 94°C to 95°C (12 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.493
Quantity 250 g
UN Number UN1760
Beilstein 1721388
Solubility Information Insoluble in water.
Formula Weight 193.05
Percent Purity 80%
Grade Technical
Chemical Name or Material Ethyl 4-bromocrotonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.