missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromocrotonate, 75%, tech.

Product Code. 10752895 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752895 100 g Glass bottle 100g
10371574 5 g Glass bottle 5g
10408840 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752895 Supplier Thermo Scientific Acros Supplier No. 118121000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37746-78-4 | C6H9BrO2 | 193.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37746-78-4
Molecular Formula C6H9BrO2
Molecular Weight (g/mol) 193.04
MDL Number MFCD00000247
InChI Key FHGRPBSDPBRTLS-ONEGZZNKSA-N
Synonym ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate
PubChem CID 5373944
IUPAC Name ethyl (E)-4-bromobut-2-enoate
SMILES CCOC(=O)C=CCBr

Specifications

Color Yellow to Brown
Density 1.4000g/mL
Boiling Point 94°C to 95°C (12.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH=CHCO2C2H5
Refractive Index 1.4900 to 1.494
Quantity 100 g
Specific Gravity 1.4
Formula Weight 193.04
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromocrotonate, tech., 75%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-bromocrotonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.