missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Bromobutanoate, TRC

Product Code. 30385906
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385906 25 g -
30375667 50 g 50g
30382082 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385906 Supplier Toronto Research Chemicals Supplier No. TRCE90029025G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.05
Synonym 1-Bromo-3-(carboethoxy)propane, 3-(Ethoxycarbonyl)propyl Bromide, 4-Bromobutanoic Acid Ethyl Ester, 4-Bromobutyric Acid Ethyl Ester, Ethyl 4-Bromobutyrate, Ethyl gamma-Bromobutyrate, NSC 133462, gamma-Bromobutyric Acid Ethyl Ester
SMILES CCOC(=O)CCCBr

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H11BrO2/c1-2-9-6(8)4-3-5-7/h2-5H2,1H3
Formula Weight 193.9942
Chemical Name or Material Ethyl 4-Bromobutanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.