missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (4-bromobenzoyl)acetate, 95%

Product Code. 10122904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122904 1 g Glass bottle 1g
10597814 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10122904 Supplier Thermo Scientific Acros Supplier No. 399160010
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 26510-95-2 | C11H11BrO3 | 271.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26510-95-2
Molecular Formula C11H11BrO3
Molecular Weight (g/mol) 271.1
MDL Number MFCD00231243
InChI Key PBDYXCKRDRCJDC-UHFFFAOYSA-N
Synonym ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester
PubChem CID 2757149
IUPAC Name ethyl 3-(4-bromophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)Br

Specifications

Color Yellow
Density 1.4320g/mL
Boiling Point 268°C to 269°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4C(O)CH2CO2CH2CH3
Quantity 1 g
Beilstein 10, I, 324
Specific Gravity 1.432
Formula Weight 271.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl (4-bromobenzoyl)acetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl (4-bromobenzoyl)acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.