missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromo-3-ethoxy-2-butenoate, tech. 90%, stab. with sodium carbonate

Product Code. 11377016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377016 1 g 1g
11387016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377016 Supplier Thermo Scientific Alfa Aesar Supplier No. L06849.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1116-50-3 | C8H13BrO3 | 237.09 g/mol

Ethyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, bases and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-50-3
Molecular Formula C8H13BrO3
Molecular Weight (g/mol) 237.09
MDL Number MFCD00051757
InChI Key FYYMWILFRUUBCX-UHFFFAOYSA-N
Synonym ethyl 4-bromo-3-ethoxybut-2-enoate, ethyl 4-bromo-3-ethoxy-2-butenoate, ethyl 2e-4-bromo-3-ethoxybut-2-enoate, ethyl e-4-bromo-3-ethoxybut-2-enoate
PubChem CID 6376123
SMILES CCOC(=O)C=C(CBr)OCC

Specifications

Boiling Point 88°C to 90°C (1 mmHg)
Odor Pungent
Refractive Index 1.496
Quantity 1 g
UN Number UN3265
Beilstein 2042896
Sensitivity Light and moisture sensitive
Solubility Information Not miscible in water.
IUPAC Name ethyl 4-bromo-3-ethoxybut-2-enoate
Formula Weight 237.1
Percent Purity 90%
Grade Technical
Chemical Name or Material Ethyl 4-bromo-3-ethoxy-2-butenoate, stabilized with sodium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.