missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-[beta-(5-Methylpyrazine-2-carboxamido)ethyl]benzene Sulfonamide Carbamate, TRC

Product Code. 30375196
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375196 Supplier Toronto Research Chemicals Supplier No. TRCE92511325MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-(beta-(5-Methylpyrazine-2-carboxyamido)ethyl)benzene Sulfonamide Carbamate
CAS 1076198-09-8
Molecular Formula C17 H20 N4 O5 S
Quantity 25 mg
Synonym Carbamic acid, N-[[4-[2-[[(5-methyl-2-pyrazinyl)carbonyl]amino]ethyl]phenyl]sulfonyl]-, ethyl ester, Ethyl [4-[2-(5-Methylpyrazine-2-carboxamido)ethyl]benzene-1-sulfonyl]carbamate
InChI Formula InChI=1S/C17H20N4O5S/c1-3-26-17(23)21-27(24,25)14-6-4-13(5-7-14)8-9-18-16(22)15-11-19-12(2)10-20-15/h4-7,10-11H,3,8-9H2,1-2H3,(H,18,22)(H,21,23)
SMILES CCOC(=O)NS(=O)(=O)c1ccc(CCNC(=O)c2cnc(C)cn2)cc1
IUPAC Name ethyl N-[4-[2-[(5-methylpyrazine-2-carbonyl)amino]ethyl]phenyl]sulfonylcarbamate
Molecular Weight (g/mol) 392.43
Formula Weight 392.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.