missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminopiperidine-1-carboxylate, 98%

Product Code. 11372797
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372797 5 g 5g
11382797 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372797 Supplier Thermo Scientific Alfa Aesar Supplier No. L15866.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 58859-46-4 | C8H16N2O2 | 172.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58859-46-4
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.228
MDL Number MFCD00006484
InChI Key GQQQULCEHJQUJT-UHFFFAOYSA-N
Synonym ethyl 4-amino-1-piperidinecarboxylate, 4-amino-1-carbethoxypiperidine, unii-54815srp4z, 1-piperidinecarboxylic acid, 4-amino-, ethyl ester, ethyl 4-aminopiperidinecarboxylate, ethyl-4-amino-1-piperidine carboxylate, 4-amino-1-piperidinecarboxylic acid ethyl ester, ethyl-4-aminopiperidin-1-carboxylat, pubchem2064, acmc-209m7k
PubChem CID 100867
IUPAC Name ethyl 4-aminopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)N

Specifications

Density 1.004
Boiling Point 118°C to 120°C (6 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.483
Quantity 5 g
Beilstein 388597
Sensitivity Air Sensitive
Formula Weight 172.23
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminopiperidine-1-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.