missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminobutyrate hydrochloride, 98%, Thermo Scientific™

Product Code. 11397016
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11397016 Supplier Thermo Scientific Alfa Aesar Supplier No. L06863.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6937-16-2
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00012915
InChI Key CXVQSUBJMYZELD-UHFFFAOYSA-N
Synonym ethyl 4-aminobutyrate hydrochloride, ethyl 4-aminobutanoate hydrochloride, 4-aminobutyric acid ethyl ester hydrochloride, ethyl 4-aminobutyrate hcl, butanoic acid, 4-amino-, ethyl ester, hydrochloride, 4-aminobutanoic acid ethyl ester hydrochloride, ethyl 4-aminobutanoate, chloride, 4-ethoxy-4-oxobutyl azanium chloride, h-gaba-oet.hcl, wln: z3vo2 &gh
PubChem CID 12649661
IUPAC Name ethyl 4-aminobutanoate;hydrochloride
SMILES CCOC(=O)CCCN.Cl

Specifications

Melting Point ∼70°C to 75°C
Linear Formula H2N(CH2)3CO2CH2CH3·HCl
Beilstein 3911780
Quantity 5 g
Formula Weight 167.64
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminobutyrate hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.