missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetylbutyrate, 97%

Product Code. 11314795
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314795 5 g 5g
11324795 25 g 25g
11334795 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314795 Supplier Thermo Scientific Alfa Aesar Supplier No. L17848.06
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13984-57-1 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13984-57-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009213
InChI Key MGPSIDGTLFKDEY-UHFFFAOYSA-N
Synonym ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa
PubChem CID 84130
IUPAC Name ethyl 5-oxohexanoate
SMILES CCOC(=O)CCCC(=O)C

Specifications

Density 0.989
Boiling Point 65°C (0.76 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.4275
Quantity 5 g
Beilstein 1762625
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material Ethyl 4-acetylbutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.