missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Acetoxybutanoate, TRC

Product Code. 30381713
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381713 250 g
30376328 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381713 Supplier Toronto Research Chemicals Supplier No. TRCE896000250G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25560-91-2
Molecular Formula C8 H14 O4
Molecular Weight (g/mol) 174.19
IUPAC Name ethyl 4-acetyloxybutanoate
SMILES CCOC(=O)CCCOC(=O)C

Specifications

Quantity 250 g
InChI Formula InChI=1S/C8H14O4/c1-3-11-8(10)5-4-6-12-7(2)9/h3-6H2,1-2H3
Formula Weight 174.0892
Chemical Name or Material Ethyl 4-Acetoxybutanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.