missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(8-chloro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate, TRC

Product Code. 30386589
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386589 10 mg -
30384407 2.5 mg 2.5mg
30373034 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386589 Supplier Toronto Research Chemicals Supplier No. TRCE90277510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133330-56-0
Molecular Formula C22 H21 Cl N2 O2
Molecular Weight (g/mol) 380.87
Synonym 1-Piperidinecarboxylic acid, 4-(8-chloro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-, ethyl ester, 11H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 1-piperidinecarboxylic acid deriv., Ethyl 4-(8-chloro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate
IUPAC Name ethyl 4-(8-chlorobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3C=Cc4cccnc24)CC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H21ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-8,11,14H,2,9-10,12-13H2,1H3
Formula Weight 380.1292
Chemical Name or Material Ethyl 4-(8-chloro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.