missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate

Product Code. 11395389
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11395389 250 mg 250mg
11305399 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11395389

Brand: Alfa Aesar™ H50397.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 333435-04-4
Molecular Formula C14H11ClO4
Molecular Weight (g/mol) 278.688
MDL Number MFCD02258008
InChI Key OPOPFTSILPZGEL-UHFFFAOYSA-N
Synonym ethyl 4-5-chlorocarbonyl-2-furyl benzoate, 4-5-chlorocarbonyl-furan-2-yl-benzoic acid ethyl ester, ethyl 4-5-chlorocarbonyl furan-2-yl benzoate, ethyl 4-5-carbonochloridoyl furan-2-yl benzoate, acmc-20amvh, ethyl 4-5-carbonochloridoylfuran-2-yl benzoate, 4-5-chlorocarbonylfuran-2-yl benzoic acid ethyl ester, benzoic acid,4-5-chlorocarbonyl-2-furanyl-, ethyl ester
PubChem CID 3131522
IUPAC Name ethyl 4-(5-carbonochloridoylfuran-2-yl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C2=CC=C(O2)C(=O)Cl

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 278.69
Chemical Name or Material Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.