missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,5-Bis(2-methoxyethoxy)-2-nitrobenzoate, TRC

Product Code. 30385709
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385709 Supplier Toronto Research Chemicals Supplier No. TRCE9000805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4,5-Bis(2-methoxyethoxy)-2-nitrobenzoate
CAS 179688-26-7
Molecular Formula C15H21NO8
Quantity 5 g
Synonym 4,5-Bis(2-methoxyethoxy)-2-nitro-benzoic Acid Ethyl Ester
InChI Formula InChI=1S/C15H21NO8/c1-4-22-15(17)11-9-13(23-7-5-20-2)14(24-8-6-21-3)10-12(11)16(18)19/h9-10H,4-8H2,1-3H3
SMILES CCOC(=O)c1cc(OCCOC)c(OCCOC)cc1[N+](=O)[O-]
Molecular Weight (g/mol) 343.33
Formula Weight 343.1267
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.