missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluorocrotonate, 98%

Product Code. 11328345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328345 1 g 1g
11338345 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328345 Supplier Thermo Scientific Alfa Aesar Supplier No. L10285.03
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25597-16-4 | C6H7F3O2 | 168.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25597-16-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00009903
InChI Key ZKRJCMKLCDWROR-ONEGZZNKSA-N
Synonym ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e
PubChem CID 5371261
IUPAC Name ethyl (E)-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)\C=C\C(F)(F)F

Specifications

Density 1.125
Boiling Point 113°C to 115°C
Flash Point 25°C (77°F)
Refractive Index 1.3601
Quantity 1 g
UN Number UN3272
Beilstein 1707262
Formula Weight 168.12
Percent Purity 98%
Chemical Name or Material Ethyl 4,4,4-trifluorocrotonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.