missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluorobutyrate, 98%

Product Code. 11307756
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307756 1 g 1g
11317756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307756 Supplier Thermo Scientific Alfa Aesar Supplier No. L11399.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 371-26-6 | C6H9F3O2 | 170.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-26-6
Molecular Formula C6H9F3O2
Molecular Weight (g/mol) 170.131
MDL Number MFCD00041398
InChI Key PSRZMXNNQTWAGB-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluorobutyrate, ethyl4,4,4-trifluorobutyrate, 4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-butyric acid ethyl ester, butanoic acid, 4,4,4-trifluoro-, ethyl ester, acmc-1cncd, ethyl-4,4,4-trifluorobutyrate, ethyl 3-trifluoromethylpropionate, butanoic acid,4,4,4-trifluoro-, ethyl ester
PubChem CID 2733273
IUPAC Name ethyl 4,4,4-trifluorobutanoate
SMILES CCOC(=O)CCC(F)(F)F

Specifications

Density 1.16
Boiling Point 126°C to 127°C
Flash Point 33°C (91°F)
Refractive Index 1.352
Quantity 1 g
UN Number UN3272
Beilstein 1769215
Formula Weight 170.13
Percent Purity 98%
Chemical Name or Material Ethyl 4,4,4-trifluorobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.