missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoroacetoacetate, 97%

Product Code. 10232892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10232892 100 mL Glass bottle 100mL
10235010 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10232892 Supplier Thermo Scientific Acros Supplier No. 119031000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-31-6 | C6H7F3O3 | 184.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-31-6
Molecular Formula C6H7F3O3
Molecular Weight (g/mol) 184.11
MDL Number MFCD00000424
InChI Key OCJKUQIPRNZDTK-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
PubChem CID 67793
IUPAC Name ethyl 4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)C(F)(F)F

Specifications

Melting Point -39.0°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 131.0°C (709.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3COCH2CO2C2H5
Refractive Index 1.3750 to 1.3780
Quantity 100 mL
Beilstein 03, II, 425
Specific Gravity 1.25
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: miscible with ethanol and ether
Formula Weight 184.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4, 4, 4-trifluoroacetoacetate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4,4,4-trifluoroacetoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.