missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(4-(3-methyl-1-phenyl-1H-Pyrazol-5-yl)piperazin-1-yl)pyrrolidine-2-carboxylate, TRC

Product Code. 30385872
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385872 250 mg 250mg
30374989 50 mg 50mg
30372887 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385872 Supplier Toronto Research Chemicals Supplier No. TRCE679025250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-(4-(3-methyl-1-phenyl-1H-Pyrazol-5-yl)piperazin-1-yl)pyrrolidine-2-carboxylate
Analyte or Component Names Ethyl 4-(4-(3-methyl-1-phenyl-1H-Pyrazol-5-yl)piperazin-1-yl)pyrrolidine-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C21 H29 N5 O2
InChI Formula InChI=1S/C21H29N5O2/c1-3-28-21(27)19-14-18(15-22-19)24-9-11-25(12-10-24)20-13-16(2)23-26(20)17-7-5-4-6-8-17/h4-8,13,18-19,22H,3,9-12,14-15H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)C1CC(CN1)N2CCN(CC2)c3cc(C)nn3c4ccccc4
IUPAC Name ethyl 4-[4-(5-methyl-2-phenylpyrazol-3-yl)piperazin-1-yl]pyrrolidine-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 383.487
Quantity 250 mg
Formula Weight 383.232
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.