missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(trimethylsilyl)propiolate, 99%, Thermo Scientific™

Product Code. 10423755 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423755 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10423755 Supplier Thermo Scientific Acros Supplier No. 381550010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16205-84-8
MDL Number MFCD00074886
InChI Key QRBKPHYAIRLCLK-UHFFFAOYSA-N
PubChem CID 568601
IUPAC Name ethyl 3-trimethylsilylprop-2-ynoate
SMILES CCOC(=O)C#C[Si](C)(C)C

Specifications

Color Colorless
Density 0.8970g/mL
Boiling Point 92.0°C to 96.0°C (28.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O2Si
Linear Formula (CH3)3SiC≡CCOOCH2CH3
Refractive Index 1.4410 to 1.4430
Quantity 1 g
Specific Gravity 0.897
Synonym ethyl 3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propynoate, ethyl 3-trimethylsilyl prop-2-ynoate, 2-propynoic acid, 3-trimethylsilyl-, ethyl ester, ethyl 3-trimethylsilyl-2-propynoate, acmc-1bvr4, ethyl 3-trimethylsily propynoate, ethyl3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propalate, trimethylsilylpropiolic acid ethyl ester
Molecular Weight (g/mol) 170.28
Formula Weight 170.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 3-(trimethylsilyl)propiolate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-(trimethylsilyl)propiolate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.