missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(trimethylsilyl)propiolate, 98%

Product Code. 15430428
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430428 5 g 5g
15440428 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430428 Supplier Thermo Scientific Alfa Aesar Supplier No. H53517.06
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 16205-84-8 | C8H14O2Si | 170.283 g/mol

Ethyl 3-(trimethylsilyl)propiolate was used in the preparation of 4-ethoxycarbonyl-3-(trimethylsilyl)furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(trimethylsilyl)propiolate was used in the preparation of 4-ethoxycarbonyl-3-(trimethylsilyl)furan.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16205-84-8
Molecular Formula C8H14O2Si
Molecular Weight (g/mol) 170.283
MDL Number MFCD00074886
InChI Key QRBKPHYAIRLCLK-UHFFFAOYSA-N
Synonym ethyl 3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propynoate, ethyl 3-trimethylsilyl prop-2-ynoate, 2-propynoic acid, 3-trimethylsilyl-, ethyl ester, ethyl 3-trimethylsilyl-2-propynoate, acmc-1bvr4, ethyl 3-trimethylsily propynoate, ethyl3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propalate, trimethylsilylpropiolic acid ethyl ester
PubChem CID 568601
IUPAC Name ethyl 3-trimethylsilylprop-2-ynoate
SMILES CCOC(=O)C#C[Si](C)(C)C

Specifications

Boiling Point 92°C to 96°C (28mmHg)
Quantity 5 g
Linear Formula (CH3)3SiC≡CCO2CH2CH3
Beilstein 1767850
Flash Point 71°C (160°F)
Solubility Information Sparingly soluble in water.
Formula Weight 170.28
Percent Purity 98%
Refractive Index 1.441
Density 0.897
Chemical Name or Material Ethyl 3-(trimethylsilyl)propiolate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.