missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-[(Tetrahydropyranyl)oxy]butanoate, TRC

Product Code. 30373422
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373422 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373422 Supplier Toronto Research Chemicals Supplier No. TRCE926483500MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-[(Tetrahydropyranyl)oxy]butanoate
CAS 104372-23-8
Molecular Formula C5 H5 N O3 S
Quantity 500 mg
Synonym 3-[(Tetrahydro-2H-pyran-2-yl)oxy]-butanoic Acid Ethyl Ester, Ethyl 3-((Tetrahydro-2H-pyran-2-yl)oxy)butanoate
InChI Formula InChI=1S/C5H5NO3S/c7-10(8,9)5-2-1-3-6-4-5/h1-4H,(H,7,8,9)
SMILES OS(=O)(=O)c1cccnc1
IUPAC Name pyridine-3-sulfonic acid
Molecular Weight (g/mol) 159.163
Formula Weight 158.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.