missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-pyridylacetate, 99%

Product Code. 10697642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697642 25 g Glass bottle 25g
10249563 5 g Glass bottle 5g
10336510 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697642 Supplier Thermo Scientific Acros Supplier No. 118920250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39931-77-6 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39931-77-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00006411
InChI Key RPWXYCRIAGBAGY-UHFFFAOYSA-N
Synonym ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate
PubChem CID 96490
IUPAC Name ethyl 2-pyridin-3-ylacetate
SMILES CCOC(=O)CC1=CN=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 80°C (0.4 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C5H4N)CH2COOC2H5
Refractive Index 1.4990 to 1.501
Quantity 25 g
Specific Gravity 1.08
Formula Weight 165.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 3-pyridylacetate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.