missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-pyridineacetate, 99%

Product Code. 11447836
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447836 25 g 25g
11437836 5 g 5g
11457836 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447836 Supplier Thermo Scientific Alfa Aesar Supplier No. A14768.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39931-77-6 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39931-77-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006411
InChI Key RPWXYCRIAGBAGY-UHFFFAOYSA-N
Synonym ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate
PubChem CID 96490
IUPAC Name ethyl 2-pyridin-3-ylacetate
SMILES CCOC(=O)CC1=CN=CC=C1

Specifications

Density 1.086
Boiling Point 79°C to 80°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5
Quantity 25 g
Beilstein 123827
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material Ethyl 3-pyridineacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.