missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific™

Product Code. 15498918
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
missing translation for 'unitSize'
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15498918

missing translation for 'mfr': Thermo Scientific Alfa Aesar H62420.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 3-oxocyclobutanecarboxylate is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87121-89-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD09834114
InChI Key BXBRFSMPBOTZHJ-UHFFFAOYSA-N
PubChem CID 22467070
IUPAC Name ethyl 3-oxocyclobutane-1-carboxylate
SMILES CCOC(=O)C1CC(=O)C1

Specifications

Quantity 250 mg
Synonym ethyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid ethyl ester, 3-oxocyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 3-oxo-, ethyl ester, 3-ethoxycarbonylcyclobutanone, ethoxycarbonyl-3-cyclobutanone, ethyl3-oxocyclobutanecarboxylate, 3-ethoxycarbonyl-1-cyclobutanone, ethyl 3-oxo-cyclobutanecarboxylate
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material Ethyl 3-oxocyclobutanecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.