missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Nitrobenzylideneacetoacetate, TRC

Product Code. 30382169
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382169 10 g
30378641 25 g
30382444 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382169 Supplier Toronto Research Chemicals Supplier No. TRCE92605010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39562-16-8
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.25
Synonym Ethyl 2-(3-Nitrobenzylidene)acetoacetate, Ethyl 2-(3'-Nitrobenzylidene)acetoacetate, Ethyl 2-(m-Nitrobenzylidene)acetoacetate, 2-[(3-Nitrophenyl)methylene]-3-oxo-Butanoic Acid Ethyl Ester
IUPAC Name ethyl (2E)-2-[(3-nitrophenyl)methylidene]-3-oxobutanoate
SMILES CCOC(=O)\C(=C\c1cccc(c1)[N+](=O)[O-])\C(=O)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H13NO5/c1-3-19-13(16)12(9(2)15)8-10-5-4-6-11(7-10)14(17)18/h4-8H,3H2,1-2H3/b12-8+
Formula Weight 263.0794
Chemical Name or Material Ethyl 3-Nitrobenzylideneacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.