missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(methylthio)propionate, 98%, Thermo Scientific™

Product Code. 11439283
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439283 500 g 500g
11429283 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11742.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13327-56-5
Molecular Formula C6H12O2S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00039900
InChI Key YSNWHRKJEKWJNY-UHFFFAOYSA-N
Synonym ethyl 3-methylthio propionate, ethyl 3-methylthio propanoate, ethyl 3-methylsulfanyl propanoate, propanoic acid, 3-methylthio-, ethyl ester, ethyl methyl mercaptopropionate, ethyl 3-methylmercaptopropionate, 3-methylthio propionic acid ethylester, 3-methylthio propanoic acid ethyl ester, unii-1avt374nii, ethyl methylthiopropanoate
PubChem CID 61592
ChEBI CHEBI:87503
IUPAC Name ethyl 3-methylsulfanylpropanoate
SMILES CCOC(=O)CCSC

Specifications

Density 1.03
Boiling Point 197°C
Flash Point 79°C (174°F)
Refractive Index 1.461
Quantity 500 g
Beilstein 1748688
Formula Weight 148.23
Percent Purity 98%
Chemical Name or Material Ethyl 3- (methylthio)propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.