missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%

Product Code. 11348045
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348045 5 g 5g
11358045 25 g 25g
11368045 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348045 Supplier Thermo Scientific Alfa Aesar Supplier No. L08218.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22232-54-8 | C7H10N2O2S | 186.229 g/mol

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

Solubility
Soluble in water, ethanol, chloroform and acetone. Sightly soluble in water.

Notes
Light sensitive. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22232-54-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD00027421
InChI Key CFOYWRHIYXMDOT-UHFFFAOYSA-N
Synonym carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
PubChem CID 31072
ChEBI CHEBI:617099
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C1=S)C

Specifications

Melting Point 122°C to 126°C
pH 5.0 to 7.0
Quantity 5 g
Beilstein 144339
Sensitivity Light sensitive
Merck Index 14,1798
Solubility Information Soluble in water,ethanol,chloroform and acetone. Sightly soluble in water.
Formula Weight 186.24
Percent Purity 97%
Chemical Name or Material Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.