missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-methyl-2-quinoxalinyl)acetate, 98%, Thermo Scientific™

Product Code. 15435857
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435857 250 mg 250mg
15425857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435857 Supplier Thermo Scientific Alfa Aesar Supplier No. H33069.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22712-18-1
Molecular Formula C13H14N2O2
Molecular Weight (g/mol) 230.267
MDL Number MFCD01820707
InChI Key ZTJBBLDJQXJLCI-UHFFFAOYSA-N
Synonym ethyl 2-3-methylquinoxalin-2-yl acetate, ethyl 3-methyl-2-quinoxalin-2-yl acetate, ethyl 3-methyl-2-quinoxalinyl acetate, ethyl 3-methyl-2-quinoxalinyl acetate #, 2-quinoxalineacetic acid, 3-methyl-, ethyl ester, quinoxaline-2-acetic acid, 3-methyl-, ethyl ester
PubChem CID 615452
IUPAC Name ethyl 2-(3-methylquinoxalin-2-yl)acetate
SMILES CCOC(=O)CC1=NC2=CC=CC=C2N=C1C

Specifications

Melting Point 59°C to 63°C
Quantity 250 mg
Formula Weight 230.27
Percent Purity 98%
Chemical Name or Material Ethyl (3-methyl-2-quinoxalinyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.